Learn More
N,O-Dinonanoylvanillylamine, Mikromol™
Product Code. 30052230
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052230 100 mg 100mg
30051311 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052230 Supplier Mikromol Supplier No. MM1033.04
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,O-Dinonanoylvanillylamine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,O-Dinonanoylvanillylamine
CAS 2727060-97-9
Analyte or Component Names N,O-Dinonanoylvanillylamine
Shipping Condition Room Temperature
Molecular Formula C26 H43 N O4
Certifications/Compliance ISO 9001
Synonym N,O-Dinonanoylvanillylamine
InChI Formula InChI=1S/C26H43NO4/c1-4-6-8-10-12-14-16-25(28)27-21-22-18-19-23(24(20-22)30-3)31-26(29)17-15-13-11-9-7-5-2/h18-20H,4-17,21H2,1-3H3,(H,27,28)
SMILES CCCCCCCCC(=O)NCc1ccc(OC(=O)CCCCCCCC)c(OC)c1
IUPAC Name [2-methoxy-4-[(nonanoylamino)methyl]phenyl] nonanoate
Molecular Weight (g/mol) 433.62
Quantity 100 mg
Formula Weight 433.3192 g/mol