Learn More
N,O-Dinonanoylvanillylamine, Mikromol™
Description
N,O-Dinonanoylvanillylamine
Specifications
Specifications
| Chemical Name or Material | N,O-Dinonanoylvanillylamine |
| CAS | 2727060-97-9 |
| Analyte or Component Names | N,O-Dinonanoylvanillylamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H43 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N,O-Dinonanoylvanillylamine |
| InChI Formula | InChI=1S/C26H43NO4/c1-4-6-8-10-12-14-16-25(28)27-21-22-18-19-23(24(20-22)30-3)31-26(29)17-15-13-11-9-7-5-2/h18-20H,4-17,21H2,1-3H3,(H,27,28) |
| SMILES | CCCCCCCCC(=O)NCc1ccc(OC(=O)CCCCCCCC)c(OC)c1 |
| IUPAC Name | [2-methoxy-4-[(nonanoylamino)methyl]phenyl] nonanoate |
| Show More |