Learn More
N,O-Diacetylbisoprolol, Mikromol™
Description
N,O-Diacetylbisoprolol
Specifications
Specifications
| Chemical Name or Material | N,O-Diacetylbisoprolol |
| CAS | 2732348-53-5 |
| Analyte or Component Names | N,O-Diacetylbisoprolol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H35 N O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N,O-Diacetylbisoprolol, [1-[[Acetyl(isopropyl)amino]methyl]-2-[4-(2-isopropoxyethoxymethyl)phenoxy]ethyl] acetate, N-[2-Acetoxy-3-[4-[[2-(1-methylethoxy)ethoxy]methyl]phenoxy]propyl]-N-(1-methylethyl)acetamide |
| InChI Formula | InChI=1S/C22H35NO6/c1-16(2)23(18(5)24)13-22(29-19(6)25)15-28-21-9-7-20(8-10-21)14-26-11-12-27-17(3)4/h7-10,16-17,22H,11-15H2,1-6H3 |
| SMILES | CC(C)OCCOCc1ccc(OCC(CN(C(C)C)C(=O)C)OC(=O)C)cc1 |
| IUPAC Name | [1-[acetyl(propan-2-yl)amino]-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-yl] acetate |
| Show More |