Learn More
N-Nitrosovarenicline, Mikromol™
Product Code. 30052258
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052258 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052258 Supplier Mikromol Supplier No. MM3380.200025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Nitrosovarenicline
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Nitrosovarenicline
CAS 2755871-02-2
Analyte or Component Names N-Nitrosovarenicline
Shipping Condition Room Temperature
Molecular Formula C13 H12 N4 O
Certifications/Compliance ISO 9001
Synonym 6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine, 7,8,9,10-tetrahydro-8-nitroso-
InChI Formula InChI=1S/C13H12N4O/c18-16-17-6-8-3-9(7-17)11-5-13-12(4-10(8)11)14-1-2-15-13/h1-2,4-5,8-9H,3,6-7H2
SMILES O=NN1CC2CC(C1)c3cc4nccnc4cc23
IUPAC Name 14-nitroso-5,8,14-triazatetracyclo[10.3.1.02,11.04,9]hexadeca-2(11),3,5,7,9-pentaene
Molecular Weight (g/mol) 240.261
Quantity 25 mg
Formula Weight 240.101 g/mol