Learn More
N-Nitrosovarenicline 0.1 mg/ml in Methanol, Mikromol
Product Code. 30463018
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30463018 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463018 Supplier Mikromol Supplier No. MM3380.20L11000
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Nitrosovarenicline 0.1 mg/ml in Methanol, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Nitrosovarenicline
CAS 2755871-02-2
Analyte or Component Names N-Nitrosovarenicline
Shipping Condition Ice pack (-20°C)
Molecular Formula C13H7D5N4O
Concentration 0.1 mg/mL
Certifications/Compliance ISO 9001
Synonym N-Nitroso Varenicline-d5
InChI Formula InChI=1S/C13H12N4O/c18-16-17-6-8-3-9(7-17)11-5-13-12(4-10(8)11)14-1-2-15-13/h1-2,4-5,8-9H,3,6-7H2/i1D,6D2,7D2
Solution Type Single Solution
SMILES [2H]C1(N(N=O)C(C2CC1C3=C2C=C4N=C(C=NC4=C3)[2H])([2H])[2H])[2H]
IUPAC Name 8-Nitroso-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline-2,7,7,9,9-d5
Solvent or Matrix Methanol
Molecular Weight (g/mol) 245.3
Quantity 1 mL
Formula Weight 245.13 g/mol