Learn More
N,N'-Dinitrosoethambutol, Mikromol
Product Code. 30461181
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461181 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461181 Supplier Mikromol Supplier No. MM3062.090025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,N'-Dinitrosoethambutol, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,N'-Dinitrosoethambutol
CAS 2792161-95-4
Analyte or Component Names N,N'-Dinitrosoethambutol
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H22 N4 O4
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C10H22N4O4/c1-3-9(7-15)13(11-17)5-6-14(12-18)10(4-2)8-16/h9-10,15-16H,3-8H2,1-2H3/t9-,10-/m0/s1
Solution Type Neat
SMILES CC[C@@H](CO)N(CCN(N=O)[C@@H](CC)CO)N=O
IUPAC Name N-[(2S)-1-hydroxybutan-2-yl]-N-[2-[[(2S)-1-hydroxybutan-2-yl]-nitrosoamino]ethyl]nitrous amide
Molecular Weight (g/mol) 262.3061
Quantity 25 mg
Formula Weight 262.1641 g/mol