Learn More
N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine Hydrogen Fumarate, Mikromol™
Description
N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine Hydrogen Fumarate
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine Hydrogen Fumarate |
| CAS | 2734096-32-1 |
| Analyte or Component Names | N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine Hydrogen Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 N2 O . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Doxylamine Hydrogen Succinate Imp. C (EP), Doxylamine Imp. C (EP), Carbinoxamine USP Related Compound C, Carbinoxamine USP RC C, N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine Hydrogen Fumarate, Doxylamine Hydrogen Succinate Imp. C (EP) as Hydrogen Fumarate, Carbinoxamine Related Compound C as Fumarate, Doxylamine Hydrogen Succinate Impurity C as Hydrogen Fumarate, Doxylamine Impurity C as Hydrogen Fumarate |
| InChI Formula | InChI=1S/C16H24ClN3.2ClH/c1-19(2)11-9-16(13-18,10-12-20(3)4)14-5-7-15(17)8-6-14;;/h5-8H,9-12H2,1-4H3;2*1H |
| SMILES | CN(C)CCOC(c1ccccc1)c2ccccn2.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;N,N-dimethyl-2-[phenyl(pyridin-2-yl)methoxy]ethanamine |
| Show More |