Learn More
N,N-Dimethyl-2-[1(RS)-1-phenyl-1-(pyridin-4-yl)ethoxy]ethanamine Hydrogen Succinate, Mikromol™
Description
N,N-Dimethyl-2-[1(RS)-1-phenyl-1-(pyridin-4-yl)ethoxy]ethanamine Hydrogen Succinate
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-2-[1(RS)-1-phenyl-1-(pyridin-4-yl)ethoxy]ethanamine Hydrogen Succinate |
| CAS | 2731671-64-8 |
| Analyte or Component Names | N,N-Dimethyl-2-[1(RS)-1-phenyl-1-(pyridin-4-yl)ethoxy]ethanamine Hydrogen Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H22 N2 O . C4 H6 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Doxylamine Hydrogen Succinate Imp. A (EP), Doxylamine Imp. A (EP), N,N-Dimethyl-2-[1(RS)-1-phenyl-1-(pyridin-4-yl)ethoxy]ethanamine Hydrogen Succinate, 4-[alpha-(2-Dimethylaminoethoxy)-alpha-methylbenzyl]-pyridine Hydrogen Succinate, Doxylamine Hydrogen Succinate Impurity A as Hydrogen Succinate, Doxylamine Impurity A as Hydrogen Succinate |
| InChI Formula | InChI=1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-7-5-4-6-8-15)16-9-11-18-12-10-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8) |
| SMILES | CN(C)CCOC(C)(c1ccccc1)c2ccncc2.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;N,N-dimethyl-2-(1-phenyl-1-pyridin-4-ylethoxy)ethanamine |
| Show More |