missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-propylethylenediamine, 97%, Thermo Scientific Chemicals

Product Code. 11371657
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371657 25 g 25g
11361657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371657 Supplier Thermo Scientific Alfa Aesar Supplier No. L04671.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14165-22-1
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00015199
InChI Key DMDXQHYISPCTGF-UHFFFAOYSA-N
Synonym n1,n1-dipropylethane-1,2-diamine, 2-di-n-propylamino ethylamine, 2-dipropylaminoethylamine, n1,n1-dipropyl-1,2-ethanediamine, 2-aminoethyl dipropylamine, n,n-dipropylethane-1,2-diamine, n,n-dipropylethylenediamine, n,n-dipropyl-1,2-ethanediamine, 1,2-ethanediamine, n,n-dipropyl, 2-di-n-propylaminoethylamine
PubChem CID 5151220
IUPAC Name N',N'-dipropylethane-1,2-diamine
SMILES CCCN(CCC)CCN

Specifications

Density 0.802
Boiling Point 186°C to 189°C
Flash Point 69°C (156°F)
Refractive Index 1.44
Quantity 25 g
UN Number UN2735
Beilstein 1734879
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N,N'-Di-n-propylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.