accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 14165-22-1

CH3NH3CNH2

CAS RN 14165-22-1

IUPAC Name: (2-aminoethyl)dipropylamine
Synonyms: (2-aminoethyl)dipropylamine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: DMDXQHYISPCTGF-UHFFFAOYSA-N
SMILES: CCCN(CCC)CCN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results