Learn More
N-Methyl-6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol Hydrochloride (N-Methylfenoldopam Hydrochloride), Mikromol™
Description
N-Methyl-6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol Hydrochloride (N-Methylfenoldopam Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | N-Methyl-6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol Hydrochloride |
| CAS | 2724762-90-5 |
| Analyte or Component Names | N-Methyl-6-Chloro-2,3,4,5-tetrahydro-1-(4-hydroxyphenyl)-1H-3-benzazepine-7,8-diol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 Cl N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6-Chloro-1-(4-hydroxyphenyl)-3-methyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol Hydrochloride, Fenoldopam Related Compound A, Fenoldopam Related Compound A as Hydrochloride |
| InChI Formula | InChI=1S/C17H18ClNO3.ClH/c1-19-7-6-12-13(8-15(21)17(22)16(12)18)14(9-19)10-2-4-11(20)5-3-10;/h2-5,8,14,20-22H,6-7,9H2,1H3;1H |
| SMILES | Cl.CN1CCc2c(Cl)c(O)c(O)cc2C(C1)c3ccc(O)cc3 |
| IUPAC Name | 6-chloro-1-(4-hydroxyphenyl)-3-methyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrochloride |
| Show More |