Learn More
N-Methyl-3,3-diphenylpropanamine Acetate, Mikromol™
Product Code. 17942476
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942476 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17942476 Supplier Mikromol Supplier No. MM3441.040025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methyl-3,3-diphenylpropanamine Acetate
TRUSTED_SUSTAINABILITY
Especificaciones
Chemical Name or Material N-Methyl-3,3-diphenylpropanamine Acetate
CAS 2727061-30-3
Analyte or Component Names N-Methyl-3,3-diphenylpropanamine Acetate
Shipping Condition Room Temperature
Molecular Formula C16 H19 N . C2 H4 O2
Certifications/Compliance ISO 17025
Synonym N-Methyl-3,3-diphenylpropanamine Acetate
InChI Formula InChI=1S/C16H19N.C2H4O2/c1-17-13-12-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;1-2(3)4/h2-11,16-17H,12-13H2,1H3;1H3,(H,3,4)
SMILES CNCCC(c1ccccc1)c2ccccc2.CC(=O)O
IUPAC Name acetic acid;N-methyl-3,3-diphenylpropan-1-amine
Molecular Weight (g/mol) 285.38
Quantity 25 mg
Formula Weight 285.1729 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.