Learn More
N-Methyl-1H-indole-5-ethanesulfonamide (2-(1H-Indol-5-yl)-N-methylethanesulfonamide), Mikromol™
Product Code. 30049460
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049460 100 mg 100mg
30051618 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049460 Supplier Mikromol Supplier No. MM0299.10
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Methyl-1H-indole-5-ethanesulfonamide (2-(1H-Indol-5-yl)-N-methylethanesulfonamide)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-1H-indole-5-ethanesulfonamide
CAS 98623-50-8
Analyte or Component Names N-Methyl-1H-indole-5-ethanesulfonamide
Shipping Condition Room Temperature
Molecular Formula C11 H14 N2 O2 S
Certifications/Compliance ISO 17025
Synonym N-Methyl-1H-indole-5-ethanesulfonamide
InChI Formula InChI=1S/C11H14N2O2S/c1-12-16(14,15)7-5-9-2-3-11-10(8-9)4-6-13-11/h2-4,6,8,12-13H,5,7H2,1H3
SMILES CNS(=O)(=O)CCc1ccc2[nH]ccc2c1
IUPAC Name 2-(1H-indol-5-yl)-N-methylethanesulfonamide
Molecular Weight (g/mol) 238.31
Quantity 100 mg
Formula Weight 238.0776 g/mol