Learn More
N-Formyl-L-leucine (3S,4R,6S)-3-Hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl Ester, Mikromol™
Product Code. 30051329
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051329 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051329 Supplier Mikromol Supplier No. MM1219.110050
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Formyl-L-leucine (3S,4R,6S)-3-Hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester
CAS 130793-27-0
Analyte or Component Names N-formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester
Shipping Condition Ice pack (-20°C)
Molecular Formula C29 H53 N O5
Certifications/Compliance ISO 9001
Synonym L-Leucine, N-formyl-, (3S,4R,6S)-3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, L-Leucine, N-formyl-, 3-hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl ester, [3S-(3α,4β,6β)]-, Orlistat Related Compound D (USP)
InChI Formula InChI=1S/C29H53NO5/c1-5-7-9-11-12-13-14-15-16-18-24-21-27(25(28(32)34-24)19-17-10-8-6-2)35-29(33)26(30-22-31)20-23(3)4/h22-27H,5-21H2,1-4H3,(H,30,31)/t24-,25-,26-,27+/m0/s1
SMILES CCCCCCCCCCC[C@H]1C[C@@H](OC(=O)[C@H](CC(C)C)NC=O)[C@H](CCCCCC)C(=O)O1
IUPAC Name [(3S,4R,6S)-3-hexyl-2-oxo-6-undecyloxan-4-yl] (2S)-2-formamido-4-methylpentanoate
Molecular Weight (g/mol) 495.73
Quantity 50 mg
Formula Weight 495.3924 g/mol