Learn More
N-Fmoc-L-homoleucine, 95%, Thermo Scientific Chemicals
Product Code. 15428818
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428818 1 g 1g
15438818 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15428818 Supplier Thermo Scientific Alfa Aesar Supplier No. H62270.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 180414-94-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00270206
InChI Key UJOQOPBFLFQOJJ-GNLPSFAGNA-N
Synonym fmoc-holeu-oh, fmoc-l-homoleucine, fmoc-hle-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-methylhexanoic acid, fmoc-homoleu-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homoleucine, fmoc-s-2-amino-5-methylhexanoic acid, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-5-methyl, fmoc-l-hol
PubChem CID 17040135
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-methylhexanoic acid
SMILES CC(C)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Specifications
Quantity 1 g
Formula Weight 367.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-homoleucine