accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3(S)OHONHOO

CAS RN 180414-94-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methylhexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-methylhexanoic acid
Molecular Weight (g/mol): 367.45
Molecular Formula: C22H25NO4
InChi Key: UJOQOPBFLFQOJJ-UHFFFAOYNA-N
SMILES: CC(C)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)