Learn More
N-Fmoc-L-glutamic acid 5-methyl ester, 97%
Product Code. 15410019
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410019 25 g 25g
15400019 5 g 5g
15420019 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15410019 Supplier Thermo Scientific Alfa Aesar Supplier No. H63358.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 145038-50-2 | C21H21NO6 | 383.40 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145038-50-2
Molecular Formula C21H21NO6
Molecular Weight (g/mol) 383.40
MDL Number MFCD01076132
InChI Key RULINAWEYRMHHQ-UHFFFAOYNA-N
Synonym fmoc-glu ome-oh, fmoc-l-glutamic acid gamma-methyl ester, fmoc-l-glu me-oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-methoxy-5-oxopentanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic acid gamma-methyl ester, n-alpha-fmoc-l-glutamic acid alpha-methyl ester, ambotzfaa1720, pubchem13156, fmoc-l-glutamic acid, a-methyl ester, n-fmoc-l-glutamic acid 5-methyl ester
PubChem CID 51340556
SMILES COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Specifications
Color White
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methoxy-5-oxopentanoic acid
Quantity 25 g
Formula Weight 383.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 5-methyl ester