accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 145038-50-2

H3COOOHONHOO

CAS RN 145038-50-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methoxy-5-oxopentanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol): 383.4
Molecular Formula: C21H21NO6
InChi Key: RULINAWEYRMHHQ-UHFFFAOYNA-N
SMILES: COC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)