missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-cyclohexyl-D-alanine, 96%, Thermo Scientific™

Product Code. 15428908
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15428908

Brand: Thermo Scientific Alfa Aesar H62382.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Fmoc-3-cyclohexyl-D-alanine is used as fine chemical intermediate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144701-25-7
Molecular Formula C24H27NO4
Molecular Weight (g/mol) 393.48
MDL Number MFCD00151912
InChI Key HIJAUEZBPWTKIV-UHFFFAOYNA-N
Synonym fmoc-d-cha-oh, fmoc-3-cyclohexyl-d-alanine, fmoc-beta-cyclohexyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-alpha-fmoc-beta-cyclohexyl-d-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylalanine, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, rarechem em wb 0095, fmoc-beta-cyclohexyl-d-ala-oh
PubChem CID 978327
SMILES OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Beilstein 8234680
IUPAC Name 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Quantity 5 g
Formula Weight 393.48
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-Fmoc-3-cyclohexyl-D-alanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.