accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 144701-25-7

OOHNHOO

CAS RN 144701-25-7

IUPAC Name: 3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-cyclohexyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 393.48
Molecular Formula: C24H27NO4
InChi Key: HIJAUEZBPWTKIV-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)