Learn More
N-Desmethyldoxylamine Hydrogen Succinate, Mikromol™
Description
N-Desmethyldoxylamine Hydrogen Succinate
Specifications
Specifications
| Chemical Name or Material | N-Desmethyldoxylamine Hydrogen Succinate |
| CAS | 2731375-61-2 |
| Analyte or Component Names | N-Desmethyldoxylamine Hydrogen Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 N2 O . C4 H6 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-Desmethyldoxylamine Hydrogen Succinate |
| InChI Formula | InChI=1S/C16H20N2O.C4H6O4/c1-16(19-13-12-17-2,14-8-4-3-5-9-14)15-10-6-7-11-18-15;5-3(6)1-2-4(7)8/h3-11,17H,12-13H2,1-2H3;1-2H2,(H,5,6)(H,7,8) |
| SMILES | CNCCOC(C)(c1ccccc1)c2ccccn2.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;N-methyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine |
| Show More |