Learn More
N-Desmethyldoxylamine Hydrogen Succinate, LoGiCal
Product Code. 30462661
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462661 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462661 Supplier LoGiCal Supplier No. LGCFOR0101.08
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Desmethyldoxylamine Hydrogen Succinate
CAS 2731375-61-2
Analyte or Component Names N-Desmethyldoxylamine Hydrogen Succinate
Shipping Condition Room Temperature
Molecular Formula C16 H20 N2 O . C4 H6 O4
Certifications/Compliance ISO 9001
Synonym N-Desmethyldoxylamine Hydrogen Succinate
InChI Formula InChI=1S/C16H20N2O.C4H6O4/c1-16(19-13-12-17-2,14-8-4-3-5-9-14)15-10-6-7-11-18-15;5-3(6)1-2-4(7)8/h3-11,17H,12-13H2,1-2H3;1-2H2,(H,5,6)(H,7,8)
Solution Type Neat
SMILES CNCCOC(C)(c1ccccc1)c2ccccn2.OC(=O)CCC(=O)O
IUPAC Name butanedioic acid;N-methyl-2-(1-phenyl-1-pyridin-2-ylethoxy)ethanamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 374.43
Quantity 10 mg
Formula Weight 374.1842 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.