Learn More
N-Boc-L-alanine N-succinimidyl ester, 98%, Thermo Scientific™
Product Code. 15405318
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15405318
Brand: Thermo Scientific Alfa Aesar H59104.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3392-05-0
Molecular Formula C12H18N2O6
Molecular Weight (g/mol) 286.284
MDL Number MFCD00065974
InChI Key COMUWNFVTWKSDT-ZETCQYMHSA-N
Synonym boc-ala-osu, succinimido s-2-tert-butoxycarbonyl amino propionate, boc-l-alanine n-succinimidyl ester, boc-l-alanine n-hydroxysuccinimide ester, s-2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino propanoate, boc-l-ala-osu, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino propanoate, boc-l-alanine n-hydroxysuccinimde ester, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-1-methyl-2-oxoethylcarbamate #, n-boc-l-alanine n-succinimidyl ester
PubChem CID 11011585
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
Specifications
Melting Point 161°C to 163°C
Quantity 1 g
Beilstein 3560476
Sensitivity Moisture sensitive
Optical Rotation −52° (c=2.5 in Dioxane)
Formula Weight 286.28
Percent Purity 98%
Chemical Name or Material N-Boc-L-alanine N-succinimidyl ester