accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3392-05-0

CH3NHOOH3CH3CCH3OONOO

CAS RN 3392-05-0

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 286.28
Molecular Formula: C12H18N2O6
InChi Key: COMUWNFVTWKSDT-UHFFFAOYNA-N
SMILES: CC(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results