missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4,4-difluoro-L-proline, 97+%, Thermo Scientific Chemicals

Product Code. p-7029575
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433597 250 mg 250mg
11406100 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433597 Supplier Thermo Scientific Alfa Aesar Supplier No. H27052.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 203866-15-3 | C10H15F2NO4 | 251.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203866-15-3
Molecular Formula C10H15F2NO4
Molecular Weight (g/mol) 251.23
MDL Number MFCD03094917
InChI Key WTMZYKCXBXPVPT-UHFFFAOYNA-N
Synonym n-boc-4,4-difluoro-l-proline, s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, n-t-boc-4,4-difluoro-l-proline, 1-tert-butoxycarbonyl-4,4-difluoro-l-proline, 2s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, boc-4,4-difluoro-l-proline, s-1-boc-4,4-difluoropyrrolidine-2-carboxylic acid, n-boc-4,4-difluoroproline
PubChem CID 22977458
SMILES CC(C)(C)OC(=O)N1CC(F)(F)CC1C(O)=O

Specifications

Melting Point 120°C to 124°C
Quantity 250 mg
IUPAC Name (2S)-4,4-difluoro-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
Formula Weight 251.24
Percent Purity ≥97%
Chemical Name or Material N-Boc-4,4-difluoro-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.