accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3H3COONOHOFF

CAS RN 203866-15-3

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 251.23
Molecular Formula: C10H15F2NO4
InChi Key: WTMZYKCXBXPVPT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(F)(F)CC1C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results