Learn More
N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
Product Code. 15455296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15455296 5 g 5g
15416268 1 g 1g
2 options
Product Code. 15455296
Brand: Thermo Scientific Alfa Aesar H59838.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate.
Solubility
Soluble in DMSO, and methanol.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5545-52-8
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00042857
InChI Key HLSLRFBLVZUVIE-LBPRGKRZSA-N
Synonym z-asp otbu-oh, n-cbz-l-aspartic acid 4-tert-butyl ester, cbz-l-aspartic acid 4-tert-butyl ester, cbz-asp obut-oh, z-asp otbu-oh.dcha, cbz-l-aspartic 4-tert-butyl ester, s-2-benzyloxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, z-l-aspartic acid 4-tert-butyl ester, 4-tert-butyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, z-asp otbu-oh.h2o
PubChem CID 111082
IUPAC Name (2S)-4-[(2-methylpropan-2-yl)oxy]-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Specifications
Linear Formula (CH3)3CO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Beilstein 2569846
Solubility Information Soluble in DMSO,and methanol.
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester