accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5545-52-8

CH3CH3CH3OO(S)HNOOOHO

CAS RN 5545-52-8

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-4-(tert-butoxy)-4-oxobutanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-4-(tert-butoxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 323.35
Molecular Formula: C16H21NO6
InChi Key: HLSLRFBLVZUVIE-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results