Learn More
N-Acetylprocainamide, Mikromol™
Description
N-Acetylprocainamide
Specifications
Specifications
| Chemical Name or Material | N-Acetylprocainamide |
| CAS | 32795-44-1 |
| Analyte or Component Names | N-Acetylprocainamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Acetylprocainamide |
| InChI Formula | InChI=1S/C15H23N3O2/c1-4-18(5-2)11-10-16-15(20)13-6-8-14(9-7-13)17-12(3)19/h6-9H,4-5,10-11H2,1-3H3,(H,16,20)(H,17,19) |
| SMILES | CCN(CC)CCNC(=O)c1ccc(NC(=O)C)cc1 |
| IUPAC Name | 4-acetamido-N-[2-(diethylamino)ethyl]benzamide |
| Show More |