Learn More
N-Acetylprocainamide, LoGiCal
Product Code. 30457575
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457575 Supplier LoGiCal Supplier No. LGCFOR0550.03
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetylprocainamide
CAS 32795-44-1
Analyte or Component Names N-Acetylprocainamide
Shipping Condition Room Temperature
Molecular Formula C15 H23 N3 O2
Certifications/Compliance ISO 9001
Synonym N-Acetylprocainamide
InChI Formula InChI=1S/C15H23N3O2/c1-4-18(5-2)11-10-16-15(20)13-6-8-14(9-7-13)17-12(3)19/h6-9H,4-5,10-11H2,1-3H3,(H,16,20)(H,17,19)
Solution Type Neat
SMILES CCN(CC)CCNC(=O)c1ccc(NC(=O)C)cc1
IUPAC Name 4-acetamido-N-[2-(diethylamino)ethyl]benzamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 277.36
Quantity 10 mg
Formula Weight 277.179 g/mol