Learn More
N-Acetylprocainamide 1.0 mg/ml in Methanol, LoGiCal
Product Code. 30458399
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458399 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458399 Supplier LoGiCal Supplier No. LGCAMP0550.0301
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetylprocainamide
CAS 32795-44-1
Analyte or Component Names N-Acetylprocainamide
Shipping Condition Room Temperature
Molecular Formula C15 H23 N3 O2
Certifications/Compliance ISO 17034
Synonym N-Acetylprocainamide
InChI Formula InChI=1S/C15H23N3O2/c1-4-18(5-2)11-10-16-15(20)13-6-8-14(9-7-13)17-12(3)19/h6-9H,4-5,10-11H2,1-3H3,(H,16,20)(H,17,19)
Solution Type Single Solution
SMILES CCN(CC)CCNC(=O)c1ccc(NC(=O)C)cc1
IUPAC Name 4-acetamido-N-[2-(diethylamino)ethyl]benzamide
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 277.36
Quantity 1 mL
Formula Weight 277.179 g/mol