missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-Thyroxine (N-Acetyllevothyroxine), Mikromol™

Product Code. 30049994
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049994 25 mg 25mg
30051473 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049994 Supplier Mikromol Supplier No. MM0694.120025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Acetyl-L-Thyroxine (N-Acetyllevothyroxine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-Thyroxine
CAS 26041-51-0
Analyte or Component Names N-Acetyl-L-Thyroxine
Shipping Condition Room Temperature
Molecular Formula C17 H13 I4 N O5
Certifications/Compliance ISO 9001
Synonym L-Tyrosine, N-acetyl-O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-, Thyroxine, N-acetyl- (6CI,7CI), Thyroxine, N-acetyl-, L- (8CI), N-Acetyl-L-thyroxine, N-Acetylthyroxine
InChI Formula InChI=1S/C17H13I4NO5/c1-7(23)22-14(17(25)26)4-8-2-12(20)16(13(21)3-8)27-9-5-10(18)15(24)11(19)6-9/h2-3,5-6,14,24H,4H2,1H3,(H,22,23)(H,25,26)/t14-/m0/s1
SMILES CC(=O)N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O
IUPAC Name (2S)-2-acetamido-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
Molecular Weight (g/mol) 818.91
Quantity 25 mg
Formula Weight 818.6972 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.