Learn More
N-[[(5-Amino-2-methylphenyl)amino]iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide, Mikromol™
Description
N-[[(5-Amino-2-methylphenyl)amino]iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide
Specifications
Specifications
| Chemical Name or Material | N-[((5-Amino-2-methylphenyl)amino)iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide |
| CAS | 2753049-11-3 |
| Analyte or Component Names | N-[((5-Amino-2-methylphenyl)amino)iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 N6 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[[(5-Amino-2-methylphenyl)amino]iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]benzamide, Benzamide, N-[[(5-amino-2-methylphenyl)amino]iminomethyl]-4-[(4-methyl-1-piperazinyl)methyl]- |
| InChI Formula | InChI=1S/C21H28N6O/c1-15-3-8-18(22)13-19(15)24-21(23)25-20(28)17-6-4-16(5-7-17)14-27-11-9-26(2)10-12-27/h3-8,13H,9-12,14,22H2,1-2H3,(H3,23,24,25,28) |
| SMILES | CN1CCN(Cc2ccc(cc2)C(=O)NC(=N)Nc3cc(N)ccc3C)CC1 |
| IUPAC Name | N-[N-(5-amino-2-methyl-phenyl)carbamimidoyl]-4-[(4-methylpiperazin-1-yl)methyl]benzamide |
| Show More |