Learn More
N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide (N-Vanillyldecanamide), Mikromol™
Product Code. 17982056
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982056 25 mg 25mg
17991196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17982056 Supplier Mikromol Supplier No. MM1033.030025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide (N-Vanillyldecanamide)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Vanillyldecanamide
CAS 31078-36-1
Analyte or Component Names N-Vanillyldecanamide
Shipping Condition Room Temperature
Molecular Formula C18 H29 N O3
Certifications/Compliance ISO 17025
Synonym N-[(4-Hydroxy-3-methoxyphenyl)methyl]-decanamide, N-Vanillyldecanamide
InChI Formula InChI=1S/C18H29NO3/c1-3-4-5-6-7-8-9-10-18(21)19-14-15-11-12-16(20)17(13-15)22-2/h11-13,20H,3-10,14H2,1-2H3,(H,19,21)
SMILES CCCCCCCCCC(=O)NCc1ccc(O)c(OC)c1
IUPAC Name N-[(4-hydroxy-3-methoxyphenyl)methyl]decanamide
Molecular Weight (g/mol) 307.43
Quantity 25 mg
Formula Weight 307.2147 g/mol