Learn More
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide, Mikromol™
Description
N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide
Specifications
Specifications
| Chemical Name or Material | N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide |
| CAS | 316373-92-9 |
| Analyte or Component Names | N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12H11F3N2O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methyl-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide |
| InChI Formula | InChI=1S/C12H11F3N2O3/c1-11(20,6-18)10(19)17-8-3-2-7(5-16)9(4-8)12(13,14)15/h2-4,18,20H,6H2,1H3,(H,17,19) |
| SMILES | CC(O)(CO)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F |
| IUPAC Name | N-[4-cyano-3-(trifluoromethyl)phenyl]-2,3-dihydroxy-2-methylpropanamide |
| Show More |