Learn More
N-[(4-chlorophenyl)methyl]cyclopentanamine, TRC
Product Code. 30040597
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040597 250 mg 250mg
30040486 500 mg 500mg
30010300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30040597 Supplier Toronto Research Chemicals Supplier No. TRCN300096250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66063-15-8
Molecular Formula C12 H16 Cl N
Molecular Weight (g/mol) 209.72
Synonym Pencycuron-PB-amine, 4-Chloro-N-cyclopentylbenzenemethanamine, N-(4-Chlorobenzyl)-N-cyclopentylamine, N-(4-Chlorobenzyl)cyclopentanamine, p-Chloro-N-cyclopentylbenzylamine, Benzenemethanamine, 4-chloro-N-cyclopentyl-
IUPAC Name N-[(4-chlorophenyl)methyl]cyclopentanamine
SMILES Clc1ccc(CNC2CCCC2)cc1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C12H16ClN/c13-11-7-5-10(6-8-11)9-14-12-3-1-2-4-12/h5-8,12,14H,1-4,9H2
Formula Weight 209.0971
Percent Purity >95%
Chemical Name or Material Pencycuron-PB-amine
Grade HPLC