CAS RN 66063-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[(4-chlorophenyl)methyl]cyclopentanamine, TRC
CAS: 66063-15-8 Molecular Formula: C12 H16 Cl N Molecular Weight (g/mol): 209.72 Synonym: Pencycuron-PB-amine,4-Chloro-N-cyclopentylbenzenemethanamine,N-(4-Chlorobenzyl)-N-cyclopentylamine,N-(4-Chlorobenzyl)cyclopentanamine,p-Chloro-N-cyclopentylbenzylamine,Benzenemethanamine, 4-chloro-N-cyclopentyl- IUPAC Name: N-[(4-chlorophenyl)methyl]cyclopentanamine SMILES: Clc1ccc(CNC2CCCC2)cc1