Learn More
N-(3-Cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine Hydrochloride, Mikromol™
Description
N-(3-Cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-(3-Cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine Hydrochloride |
| CAS | 2732347-31-6 |
| Analyte or Component Names | N-(3-Cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H33 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Alverine Citrate Imp. D (EP), Alverine Imp. D (EP), N-(3-Cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine Hydrochloride, Alverine Citrate Imp. D (EP) as Hydrochloride, Alverine Citrate Impurity D as Hydrochloride, Alverine Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C14H21NO3.ClH/c1-5-18-14(16)13(10-15(2)3)11-6-8-12(17-4)9-7-11;/h6-9,13H,5,10H2,1-4H3;1H |
| SMILES | Cl.CCN(CCCC1CCCCC1)CCCc2ccccc2 |
| IUPAC Name | N-(3-cyclohexylpropyl)-N-ethyl-3-phenylpropan-1-amine;hydrochloride |
| Show More |