Learn More
N,3,3-Trimethyl-1,5-dioxaspiro[5.5]undecan-9-amine Hydrochloride (4-Methylaminocyclohexanone 2',2'-Dimethyltrimethylene Ketal Hydrochloride), Mikromol
Click to view available options
Quantity:
25 mg
Description
N,3,3-Trimethyl-1,5-dioxaspiro[5.5]undecan-9-amine Hydrochloride (4-Methylaminocyclohexanone 2',2'-Dimethyltrimethylene Ketal Hydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | 4-(Methylamino)cyclohexanone 2,2-Dimethyltrimethylene Ketal Hydrochloride |
| CAS | 158747-10-5 |
| Analyte or Component Names | 4-(Methylamino)cyclohexanone 2,2-Dimethyltrimethylene Ketal Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12H23NO2 . HCl |
| Certifications/Compliance | ISO 9001 |
| Solution Type | Neat |
| SMILES | Cl.CNC1CCC2(CC1)OCC(C)(C)CO2 |
| Molecular Weight (g/mol) | 249.78 |
| Quantity | 25 mg |