Learn More
N-(2-Pyridinyl)oxamic Acid Hydrochloride, Mikromol™
Product Code. 17963806
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963806 25 mg 25mg
17954176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17963806 Supplier Mikromol Supplier No. MM0084.170025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Pyridinyl)oxamic Acid Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(2-Pyridinyl)oxamic Acid Hydrochloride
CAS 2731671-59-1
Analyte or Component Names N-(2-Pyridinyl)oxamic Acid Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C7 H6 N2 O3 . Cl H
Certifications/Compliance ISO 9001
Synonym Nateglinide IPP Impurity Dicyclohexylammonium Salt, N-(2-Pyridinyl)oxamic Acid Hydrochloride
InChI Formula InChI=1S/C16H24ClN3.2ClH/c1-19(2)11-9-16(13-18,10-12-20(3)4)14-5-7-15(17)8-6-14;;/h5-8H,9-12H2,1-4H3;2*1H
SMILES Cl.OC(=O)C(=O)Nc1ccccn1
IUPAC Name 2-oxo-2-(pyridin-2-ylamino)acetic acid;hydrochloride
Molecular Weight (g/mol) 202.6
Quantity 25 mg
Formula Weight 202.0145 g/mol