Learn More
N-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)-methoxy]ethyl]formamide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)-methoxy]ethyl]formamide
Specifications
Specifications
| Chemical Name or Material | N-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)-methoxy]ethyl]formamide |
| CAS | 911805-54-4 |
| Analyte or Component Names | N-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)-methoxy]ethyl]formamide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H13 Cl4 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)-methoxy]ethyl]formamide |
| InChI Formula | InChI=1S/C16H13Cl4NO2/c17-11-2-1-10(14(19)5-11)8-23-16(7-21-9-22)13-4-3-12(18)6-15(13)20/h1-6,9,16H,7-8H2,(H,21,22) |
| SMILES | Clc1ccc(COC(CNC=O)c2ccc(Cl)cc2Cl)c(Cl)c1 |
| IUPAC Name | N-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]formamide |
| Show More |