Learn More
N-(2-[[2-([4-[2-(2-Hydroxyethoxy)ethyl]piperazin-1-yl]carbonyl)phenyl]thio]phenyl)acetamide Fumarate, Mikromol™
Beschreibung
N-(2-[[2-([4-[2-(2-Hydroxyethoxy)ethyl]piperazin-1-yl]carbonyl)phenyl]thio]phenyl)acetamide Fumarate
Spezifikation
Spezifikation
| Chemical Name or Material | N-(2-[(2-([4-[2-(2-Hydroxyethoxy)ethyl]piperazin-1-yl]carbonyl)phenyl)thio]phenyl)acetamide Fumarate |
| CAS | 2734483-49-7 |
| Analyte or Component Names | N-(2-[(2-([4-[2-(2-Hydroxyethoxy)ethyl]piperazin-1-yl]carbonyl)phenyl)thio]phenyl)acetamide Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H29 N3 O4 S . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Quetiapine Fumarate Imp. K (EP), Quetiapine Imp. K (EP), Quetiapine Fumarate Imp. K (EP) as Fumarate, N-(2-[[2-([4-[2-(2-Hydroxyethoxy)ethyl]piperazin-1-yl]carbonyl)phenyl]thio]phenyl)acetamide Fumarate, Quetiapine Fumarate Impurity K as Fumarate, Quetiapine Impurity K as Fumarate |
| InChI Formula | InChI=1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-7-5-4-6-8-15)16-9-11-18-12-10-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8) |
| SMILES | CC(=O)Nc1ccccc1Sc2ccccc2C(=O)N3CCN(CCOCCO)CC3.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;N-[2-[2-[4-[2-(2-hydroxyethoxy)ethyl]piperazine-1-carbonyl]phenyl]sulfanylphenyl]acetamide |
| Mehr anzeigen |