Learn More
Macherey-Nagel™ N 12 Septa Replacement Screw Caps
4.90 € - 27.50 €
Specifications
Septum | Septa N 12 |
---|---|
Quantity | 100 Pk. |
For Use With (Equipment) | Micro Reaction Vials N 13 |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
10206551
|
Macherey-Nagel™
70260 |
PTFE Virginal |
4.90 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
12794658
|
Macherey-Nagel™
702292 |
Silicone, PTFE |
27.50 €
Pack of 100 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Specifications
Septa N 12 | |
100 Pk. | |
Micro Reaction Vials N 13 |