Learn More
Macherey-Nagel™ N 12 Septa Replacement Screw Caps
Product Code. 10206551 Shop All Macherey-Nagel Products
Change view
Click to view available options
Material:
PTFE Virginal
Silicone, PTFE
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10206551 PTFE Virginal Pack of 100
12794658 Silicone, PTFE Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10206551 Supplier Macherey-Nagel™ Supplier No. 70260
In Stock
Add to basket
Add to basket
Specifications
Color White
Material PTFE Virginal
Septum Septa N 12
Quantity 100 Pk.
Thickness (Metric) 0.25 mm
For Use With (Equipment) Micro Reaction Vials N 13