Learn More
N-(1,1-Dimethylethyl)-3-oxo-4-aza-5alpha-androstane-17beta-carboxamide (Dihydrofinasteride), Mikromol™
Description
N-(1,1-Dimethylethyl)-3-oxo-4-aza-5alpha-androstane-17beta-carboxamide (Dihydrofinasteride)
Specifications
Specifications
| Chemical Name or Material | Dihydrofinasteride |
| CAS | 98319-24-5 |
| Analyte or Component Names | Dihydrofinasteride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H38 N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-(1,1-Dimethylethyl)-3-oxo-4-aza-5alpha-androstane-17beta-carboxamide, Finasteride Imp. A (EP), Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrostane-17-carboxamide, N-(1,1-dimethylethyl)-3-oxo-, (5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-(1,1-Dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, 1,2-Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-4-aza-5α-androstan-17β-carboxylic acid N-(tert-butyl)amide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17-carboxamide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17β-carboxamide, Finasteride Impurity A |
| InChI Formula | InChI=1S/C23H38N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h14-18H,6-13H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1 |
| SMILES | CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxamide |
| Show More |