Learn More
MYH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05634-20ul
 View more versions of this product
Product Code. 18669477
  • 201.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
MYH4 Polyclonal antibody specifically detects MYH4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MYH4
Polyclonal
Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
MyHC-2B, MyHC-IIb, myosin heavy chain 4, myosin heavy chain IIb, myosin heavy chain, skeletal muscle, fetal, myosin, heavy chain 4, skeletal muscle, myosin, heavy polypeptide 4, skeletal muscle, myosin-4
A synthetic peptide corresponding to a sequence within amino acids 1500-1600 of human MYH4 (NP_060003.2). ETLKRENKNLQQEISDLTEQIAEGGKHIHELEKVKKQLDHEKSELQTSLEEAEASLEHEEGKILRIQLELNQVKSEIDRKIAEKDEELDQLKRNHLRVVES
20 μg
Cell Biology, Cytoskeleton Markers, Signal Transduction
4622
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications