missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
247.00 €
Specifications
| Antigen | MyD88 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, Immunohistochemistry 2-5 μg/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 2-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
MyD88 Polyclonal specifically detects MyD88 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MyD88 | |
| Western Blot 0.5-2 μg/mL, Immunohistochemistry 2-5 μg/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 2-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| MYD88 | |
| IgG | |
| Protein G purified | |
| Because of high sequence homology, this antibody is expected to detect multiple isoforms of MyD88 protein. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q99836 | |
| 4615 | |
| A synthetic peptide corresponding to human MyD88 (between amino acids 200-270). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title