missing translation for 'onlineSavingsMsg'
Learn More

MyD88 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 18 publications

Brand:  Novus Biologicals NB100-56698SS

 View more versions of this product

Product Code. 18550561

  • 247.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MyD88 Polyclonal specifically detects MyD88 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MyD88
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Rabbit
Protein G purified
RUO
4615
Human
Purified
Western Blot
1 mg/mL
Western Blot 0.5-2 μg/mL, Immunohistochemistry 2-5 μg/mL, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 2-5 μg/mL
Q99836
MYD88
A synthetic peptide corresponding to human MyD88 (between amino acids 200-270).
0.025 mg
Primary
Because of high sequence homology, this antibody is expected to detect multiple isoforms of MyD88 protein.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only