missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Antibody (MUM1p), Novus Biologicals™
Mouse Monoclonal Antibody
608.00 € - 1020.00 €
Specifications
| Antigen | MUM1 |
|---|---|
| Clone | MUM1p |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201925
|
Novus Biologicals
NB200-356-0.25ML |
0.25 mL |
608.00 €
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18205140
|
Novus Biologicals
NB200-356 |
0.5 mL |
1020.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUM1 Monoclonal specifically detects MUM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| MUM1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
| MUM1 | |
| Primary |
| MUM1p | |
| Monoclonal | |
| Supernatant | |
| Human | |
| 84939 | |
| Recombinant GST-MUM1 protein. | |
| Multiple myeloma oncogene-1 (MUM1) is a 50kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto