missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Antibody (MUM1p), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-356-0.25ML
This item is not returnable.
View return policy
Description
MUM1 Monoclonal specifically detects MUM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| MUM1 | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with sodium azide | |
| MUM1 | |
| Recombinant GST-MUM1 protein. | |
| Primary | |
| Multiple myeloma oncogene-1 (MUM1) is a 50kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MUM1p | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
| Mouse | |
| 0.25 mL | |
| 84939 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction